Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnase1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014223.2 | NP_001014245.2 | 5 - 6 | 688 | 86 | |
XM_006229602.1 | XP_006229664.1 | 4 - 5 | 665 | 86 | ||
GenBank mRNA | BC079074.1 | - | 5 - 6 | 644 | 7122158 | 86 |
DQ116785.1 | - | 5 - 6 | 688 | 86 | ||
FQ232580.1 | - | 4 - 5 | 614 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |