Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc17a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271214.1 | NP_001258143.1 | 9 - 10 | 1290 | 90 | |
XM_008771657.1 | XP_008769879.1 | 9 - 10 | 1295 | 90 | ||
XM_008771658.1 | XP_008769880.1 | 10 - 11 | 1609 | 90 | ||
XM_008771659.1 | XP_008769881.1 | 10 - 11 | 1679 | 90 | ||
XM_008771660.1 | XP_008769882.1 | 8 - 9 | 1061 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Transporter Rattus norvegicus Probe spans exons |