Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc17a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107353.1 | NP_001100823.1 | 5 - 6 | 941 | 93 | |
XM_006253966.2 | XP_006254028.1 | 5 - 6 | 1018 | 93 | ||
XM_006253967.2 | XP_006254029.1 | 5 - 6 | 765 | 93 | ||
XM_006253968.2 | XP_006254030.1 | 5 - 6 | 1034 | 93 | ||
XM_006253969.2 | XP_006254031.1 | 5 - 6 | 1018 | 93 | ||
XM_008771654.1 | XP_008769876.1 | 5 - 6 | 2202 | 93 | ||
XM_008771655.1 | XP_008769877.1 | 5 - 6 | 1949 | 93 | ||
XM_008771656.1 | XP_008769878.1 | 5 - 6 | 2202 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Transporter Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |