Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nde1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006245849.2 | XP_006245911.1 | 1 - 2 | 77 | 92 | |
XM_006245851.1 | XP_006245913.1 | 1 - 2 | 77 | 92 | ||
XM_008767443.1 | XP_008765665.1 | 1 - 2 | 61 | 92 | ||
XM_008767444.1 | XP_008765666.1 | 1 - 2 | 61 | 92 | ||
XM_008767445.1 | XP_008765667.1 | 1 - 2 | 61 | 92 |
Gene Symbol: LOC103690118 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006245690.2 | XP_006245752.1 | 1 - 2 | 106 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |