Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108382.1 | NP_001101852.1 | 7 - 8 | 654 | 145 | |
XM_006252132.2 | XP_006252194.1 | 7 - 8 | 653 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |