Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gata4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144730.1 | NP_653331.1 | 2 - 3 | 1169 | 92 | |
GenBank mRNA | L22761.1 | - | 2 - 3 | 1169 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |