Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nefl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031783.1 | NP_113971.1 | 2 - 3 | 1244 | 122 | |
GenBank mRNA | AF031880.1 | - | 2 - 3 | 1244 | 122 | |
M25638.1 | - | 2 - 3 | 582 | 122 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |