Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam81a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108163.1 | NP_001101633.1 | 1 - 2 | 278 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |