Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rora | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008766408.1 | XP_008764630.1 | 6 - 7 | 1353 | 113 | |
XM_008766409.1 | XP_008764631.1 | 6 - 7 | 1351 | 113 | ||
XM_008766410.1 | XP_008764632.1 | 6 - 7 | 1009 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Nuclear Receptor Rattus norvegicus Probe spans exons |