Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pygo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191117.1 | NP_001178046.1 | 2 - 3 | 135 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |