Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lilrb3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037357.2 | NP_001032434.1 | 81 | |||
XM_006228032.2 | XP_006228094.1 | 5 - 6 | 1000 | 81 | ||
XM_006228033.2 | XP_006228095.1 | 5 - 6 | 1000 | 81 | ||
XM_006228034.2 | XP_006228096.1 | 4 - 5 | 715 | 81 | ||
GenBank mRNA | AF169636.1 | - | 949 | 81 | ||
AF169637.1 | - | 6 - 7 | 958 | 81 | ||
AF169638.1 | - | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |