Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Idua | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172084.1 | NP_001165555.1 | 13 - 14 | 1904 | 77 | |
XM_008769938.1 | XP_008768160.1 | 11 - 12 | 1542 | 77 | ||
GenBank mRNA | BC083854.1 | - | 11 - 12 | 1405 | 77 | |
BC103733.1 | - | 11 - 12 | 1922 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |