Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011665.2 | NP_001011665.1 | 6 - 7 | 1104 | 152 | |
XM_008772692.1 | XP_008770914.1 | 7 - 8 | 1060 | 152 | ||
XM_008772693.1 | XP_008770915.1 | 5 - 6 | 2665 | 152 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |