Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106376.1 | NP_001099846.1 | 1 - 2 | 640 | 68 | |
XM_008772691.1 | XP_008770913.1 | 2 - 3 | 551 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |