Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Large | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108439.1 | NP_001101909.1 | 7 - 8 | 1352 | 71 | |
XM_006255140.2 | XP_006255202.1 | 7 - 8 | 2003 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |