Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152242.2 | NP_689448.2 | 3 - 4 | 773 | 68 | |
XM_006255066.2 | XP_006255128.1 | 3 - 4 | 1825 | 68 | ||
XM_008772264.1 | XP_008770486.1 | 3 - 4 | 975 | 68 | ||
XM_008772265.1 | XP_008770487.1 | 4 - 5 | 1239 | 68 | ||
XM_008772266.1 | XP_008770488.1 | 3 - 4 | 2485 | 68 | ||
XM_008772267.1 | XP_008770489.1 | 3 - 4 | 2748 | 68 | ||
XM_008772268.1 | XP_008770490.1 | 4 - 5 | 1072 | 68 | ||
XM_008772269.1 | XP_008770491.1 | 3 - 4 | 1854 | 68 | ||
XM_008772270.1 | XP_008770492.1 | 3 - 4 | 2330 | 68 | ||
GenBank mRNA | CV116515.1 | - | 1 - 2 | 229 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Rattus norvegicus Probe spans exons |