Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ciapin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007689.1 | NP_001007690.1 | 8 - 9 | 1135 | 97 | |
XM_006255106.2 | XP_006255168.1 | 8 - 9 | 1222 | 97 | ||
XM_006255107.2 | XP_006255169.1 | 8 - 9 | 934 | 97 | ||
XM_006255108.2 | XP_006255170.1 | 8 - 9 | 937 | 97 | ||
GenBank mRNA | BC083753.1 | - | 8 - 9 | 1135 | 7192604 | 97 |
FQ211546.1 | - | 8 - 9 | 919 | 97 | ||
FQ213033.1 | - | 8 - 9 | 966 | 97 | ||
FQ230885.1 | - | 8 - 9 | 1147 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons |