Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153624.2 | NP_705888.2 | 21 - 22 | 3338 | 98 | |
XM_006254403.2 | XP_006254465.1 | 21 - 22 | 3394 | 98 | ||
GenBank mRNA | BC166779.1 | - | 3 - 4 | 376 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |