Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153624.2 | NP_705888.2 | 4 - 5 | 559 | 62 | |
XM_006254403.2 | XP_006254465.1 | 4 - 5 | 618 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |