Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prr13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008379.1 | NP_001008380.1 | 3 - 4 | 456 | 70 | |
GenBank mRNA | BC086446.1 | - | 3 - 4 | 456 | 7303132 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons |