Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008828.1 | NP_001008828.1 | 2 - 3 | 684 | 64 | |
XM_006242325.2 | XP_006242387.1 | 2 - 3 | 884 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |