Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008828.1 | NP_001008828.1 | 6 - 7 | 1136 | 81 | |
XM_006242325.2 | XP_006242387.1 | 6 - 7 | 1336 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons |