Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aqp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012909.2 | NP_037041.2 | 3 - 4 | 689 | 88 | |
GenBank mRNA | BC128705.1 | - | 3 - 4 | 675 | 7109642 | 88 |
D13906.1 | - | 3 - 4 | 689 | 88 | ||
L28112.1 | - | 3 - 4 | 728 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons |