Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spats2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191614.1 | NP_001178543.1 | 4 - 5 | 417 | 84 | |
XM_006257350.1 | XP_006257412.1 | 5 - 6 | 839 | 84 | ||
XM_006257351.2 | XP_006257413.1 | 5 - 6 | 595 | 84 | ||
XM_008765701.1 | XP_008763923.1 | 5 - 6 | 2485 | 84 | ||
XM_008765702.1 | XP_008763924.1 | 5 - 6 | 896 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |