Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aaas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106795.1 | NP_001100265.1 | 3 - 4 | 487 | 97 | |
XM_006242387.2 | XP_006242449.1 | 2 - 3 | 404 | 97 | ||
XM_008765704.1 | XP_008763926.1 | 2 - 3 | 447 | 97 | ||
XM_008765705.1 | XP_008763927.1 | 2 - 3 | 428 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |