Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005545.1 | NP_001005545.1 | 5 - 6 | 767 | 81 | |
GenBank mRNA | BC082112.1 | - | 5 - 6 | 767 | 7135321 | 81 |
FQ213994.1 | - | 3 - 4 | 832 | 81 | ||
FQ230067.1 | - | 5 - 6 | 797 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |