Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scaf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271170.1 | NP_001258099.1 | 11 - 12 | 3555 | 107 | |
GenBank mRNA | AY724482.1 | - | 2 - 3 | 1968 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |