Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glycam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012794.1 | NP_036926.1 | 3 - 4 | 336 | 116 | |
GenBank mRNA | L08100.1 | - | 3 - 4 | 336 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |