Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012633.2 | NP_036765.2 | 7 - 8 | 1310 | 155 | |
XM_006257321.2 | XP_006257383.1 | 6 - 7 | 2189 | 155 | ||
XM_006257322.2 | XP_006257384.1 | 7 - 8 | 2091 | 155 | ||
XM_008765669.1 | XP_008763891.1 | 6 - 7 | 2191 | 155 | ||
GenBank mRNA | AF031878.1 | - | 7 - 8 | 1303 | 155 | |
BC100656.1 | - | 7 - 8 | 1310 | 7936222 | 155 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |