Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfe2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012224.1 | NP_001012224.1 | 2 - 3 | 422 | 66 | |
XM_006242431.2 | XP_006242493.1 | 3 - 4 | 634 | 66 | ||
XM_006242432.2 | XP_006242494.1 | 2 - 3 | 453 | 66 | ||
XM_006242433.2 | XP_006242495.1 | 2 - 3 | 1079 | 66 | ||
GenBank mRNA | BC078925.1 | - | 2 - 3 | 422 | 7106450 | 66 |
FQ233359.1 | - | 2 - 3 | 256 | 66 | ||
FQ234994.1 | - | 2 - 3 | 267 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |