Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vdr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017058.1 | NP_058754.1 | 5 - 6 | 563 | 80 | |
GenBank mRNA | J04147.1 | - | 5 - 6 | 563 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Nuclear Receptor Detect Genomic DNA Rattus norvegicus Probe spans exons |