Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Reg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004096.1 | NP_001004096.1 | 1 - 2 | 66 | 103 | |
XM_006233009.2 | XP_006233071.1 | 2 - 3 | 269 | 103 | ||
XM_008761321.1 | XP_008759543.1 | 2 - 3 | 232 | 103 | ||
GenBank mRNA | AB164049.1 | - | 1 - 2 | 66 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |