Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Reg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004096.1 | NP_001004096.1 | 3 - 4 | 300 | 56 | |
XM_006233009.2 | XP_006233071.1 | 4 - 5 | 503 | 56 | ||
XM_008761321.1 | XP_008759543.1 | 4 - 5 | 466 | 56 | ||
GenBank mRNA | AB164049.1 | - | 3 - 4 | 300 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |