Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107694.1 | NP_001101164.1 | 7 - 8 | 602 | 129 | |
XM_006232876.2 | XP_006232938.1 | 6 - 7 | 2100 | 129 | ||
XM_006232878.2 | XP_006232940.1 | 6 - 7 | 2409 | 129 | ||
XM_008761300.1 | XP_008759522.1 | 5 - 6 | 851 | 129 | ||
XM_008761301.1 | XP_008759523.1 | 6 - 7 | 2355 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |