Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031645.1 | NP_001026815.1 | 1 - 2 | 201 | 83 | |
GenBank mRNA | BC099131.1 | - | 1 - 2 | 201 | 7444695 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |