Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031645.1 | NP_001026815.1 | 5 - 6 | 624 | 66 | |
GenBank mRNA | BC099131.1 | - | 5 - 6 | 624 | 7444695 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |