Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109340.1 | NP_001102810.1 | 6 - 7 | 743 | 60 | |
XM_008761327.1 | XP_008759549.1 | 6 - 7 | 2568 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons |