Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lingo4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109189.1 | NP_001102659.1 | 1 - 2 | 190 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Rattus norvegicus Probe spans exons |