Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbx15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106451.1 | NP_001099921.1 | 2 - 3 | 317 | 129 | |
XM_006233027.2 | XP_006233089.1 | 3 - 4 | 892 | 129 | ||
XM_008761285.1 | XP_008759507.1 | 3 - 4 | 5191 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |