Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Them4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025017.1 | NP_001020188.1 | 2 - 3 | 329 | 89 | |
GenBank mRNA | BC093385.1 | - | 2 - 3 | 329 | 7389383 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |