Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sf3b4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011951.1 | NP_001011951.1 | 4 - 5 | 1152 | 75 | |
GenBank mRNA | BC078997.1 | - | 4 - 5 | 1152 | 7112738 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons |