Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nhlh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006233047.2 | XP_006233109.1 | 2 - 3 | 531 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Rattus norvegicus Probe spans exons |