Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lysmd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024302.1 | NP_001019473.1 | 1 - 2 | 659 | 82 | |
GenBank mRNA | BC089113.1 | - | 1 - 2 | 659 | 7319992 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |