Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam19a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006233102.2 | XP_006233164.1 | 3 - 4 | 648 | 73 | |
XM_006233103.2 | XP_006233165.1 | 4 - 5 | 1607 | 73 | ||
XM_008761434.1 | XP_008759656.1 | 5 - 6 | 1206 | 73 | ||
XM_008761435.1 | XP_008759657.1 | 3 - 4 | 672 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Probe spans exons |