Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmbox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006252219.2 | XP_006252281.1 | 5 - 6 | 1026 | 114 | |
XM_006252220.2 | XP_006252282.1 | 5 - 6 | 1026 | 114 | ||
XM_006252221.2 | XP_006252283.1 | 5 - 6 | 1025 | 114 | ||
XM_008770802.1 | XP_008769024.1 | 5 - 6 | 953 | 114 | ||
XM_008770803.1 | XP_008769025.1 | 5 - 6 | 1026 | 114 | ||
XM_008770804.1 | XP_008769026.1 | 5 - 6 | 1026 | 114 | ||
XM_008770805.1 | XP_008769027.1 | 5 - 6 | 1026 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |