Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrna2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133420.1 | NP_596911.1 | 4 - 5 | 573 | 96 | |
XM_008770735.1 | XP_008768957.1 | 4 - 5 | 415 | 96 | ||
GenBank mRNA | AY574253.1 | - | 3 - 4 | 269 | 96 | |
L10077.1 | - | 4 - 5 | 573 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Rattus norvegicus Probe spans exons |