Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053021.2 | NP_444180.2 | 4 - 5 | 541 | 77 | |
XM_006252094.2 | XP_006252156.1 | 3 - 4 | 419 | 77 | ||
GenBank mRNA | BC061534.1 | - | 3 - 4 | 320 | 6886725 | 77 |
FQ209467.1 | - | 3 - 4 | 317 | 77 | ||
FQ209498.1 | - | 3 - 4 | 318 | 77 | ||
FQ209623.1 | - | 3 - 4 | 316 | 77 | ||
FQ213433.1 | - | 3 - 4 | 315 | 77 | ||
FQ218237.1 | - | 3 - 4 | 316 | 77 | ||
FQ218812.1 | - | 3 - 4 | 316 | 77 | ||
FQ219030.1 | - | 3 - 4 | 316 | 77 | ||
M16975.1 | - | 4 - 5 | 541 | 77 | ||
M64723.1 | - | 3 - 4 | 306 | 77 | ||
X13231.1 | - | 3 - 4 | 305 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Rattus norvegicus Probe spans exons |