Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpysl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105717.2 | NP_001099187.1 | 6 - 7 | 845 | 72 | |
XM_006252097.2 | XP_006252159.1 | 6 - 7 | 1076 | 72 | ||
GenBank mRNA | CB729753.1 | - | 3 - 4 | 251 | 72 | |
Z46882.1 | - | 6 - 7 | 798 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |