Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fzd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153474.1 | NP_703204.1 | 1 - 2 | 49 | 132 | |
GenBank mRNA | AF323956.1 | - | 1 - 2 | 49 | 132 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Rattus norvegicus Probe spans exons |