Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sacs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006252167.2 | XP_006252229.1 | 11 | 15354 | 143 | |
XM_006252168.2 | XP_006252230.1 | 10 | 14882 | 143 | ||
XM_006252169.2 | XP_006252231.1 | 10 | 15327 | 143 | ||
XM_008770792.1 | XP_008769014.1 | 10 | 15207 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Both primers and probe map within a single exon |